Assay Details
| Gene Symbol: PUR ALPHA-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128768.3 | NP_565736.1 | 1 - 2 | 439 | 87 | |
| NM_179851.3 | NP_850182.1 | 1 - 2 | 694 | 87 | ||
| GenBank mRNA | AF136152.1 | - | 1 - 2 | 435 | 87 | |
| AF419556.1 | - | 1 - 2 | 422 | 87 | ||
| AF446882.1 | - | 1 - 2 | 309 | 87 | ||
| AY052714.1 | - | 1 - 2 | 423 | 87 | ||
More Information
Set Membership: |
|

PUR ALPHA-1
Genomic Map