Assay Details
| Gene Symbol: PUR ALPHA-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128768.3 | NP_565736.1 | 2 - 3 | 488 | 130 | |
| NM_179851.3 | NP_850182.1 | 2 - 3 | 743 | 130 | ||
| GenBank mRNA | AF136152.1 | - | 2 - 3 | 484 | 130 | |
| AF419556.1 | - | 2 - 3 | 471 | 130 | ||
| AF446882.1 | - | 2 - 3 | 358 | 130 | ||
| AY052714.1 | - | 2 - 3 | 472 | 130 | ||
More Information
Set Membership: |
|

PUR ALPHA-1
Genomic Map