Assay Details
| Gene Symbol: BAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128788.3 | NP_180788.2 | 1 - 2 | 277 | 93 | |
| GenBank mRNA | AY136463.1 | - | 1 - 2 | 204 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BAM6| Gene Symbol: BAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128788.3 | NP_180788.2 | 1 - 2 | 277 | 93 | |
| GenBank mRNA | AY136463.1 | - | 1 - 2 | 204 | 93 | |
Set Membership: |
|