Assay Details
| Gene Symbol: LUH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036389.4 | NP_001031466.2 | 18 - 19 | 2491 | 91 | |
| NM_001202730.1 | NP_001189659.1 | 18 - 19 | 2542 | 91 | ||
| NM_128829.4 | NP_565749.1 | 18 - 19 | 2497 | 91 | ||
| NM_179861.3 | NP_850192.1 | 18 - 19 | 2443 | 91 | ||
| NM_179862.2 | NP_850193.1 | 17 - 18 | 2572 | 91 | ||
| NM_179863.3 | NP_850194.1 | 18 - 19 | 2539 | 91 | ||
| NM_179864.3 | NP_850195.2 | 18 - 19 | 2388 | 91 | ||
| GenBank mRNA | AF367306.1 | - | 18 - 19 | 2485 | 91 | |
| BT002219.1 | - | 16 - 17 | 2218 | 91 | ||
More Information
Set Membership: |
|

LUH
Genomic Map