Assay Details
| Gene Symbol: LUH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036389.4 | NP_001031466.2 | 6 - 7 | 801 | 71 | |
| NM_001202730.1 | NP_001189659.1 | 6 - 7 | 789 | 71 | ||
| NM_128829.4 | NP_565749.1 | 6 - 7 | 801 | 71 | ||
| NM_179861.3 | NP_850192.1 | 6 - 7 | 747 | 71 | ||
| NM_179862.2 | NP_850193.1 | 5 - 6 | 876 | 71 | ||
| NM_179863.3 | NP_850194.1 | 6 - 7 | 843 | 71 | ||
| NM_179864.3 | NP_850195.2 | 6 - 7 | 692 | 71 | ||
| GenBank mRNA | AF367306.1 | - | 6 - 7 | 789 | 71 | |
| BT002219.1 | - | 4 - 5 | 522 | 71 | ||
More Information
Set Membership: |
|

LUH
Genomic Map