Assay Details
| Gene Symbol: LUH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036389.4 | NP_001031466.2 | 8 - 9 | 982 | 78 | |
| NM_128829.4 | NP_565749.1 | 8 - 9 | 982 | 78 | ||
| NM_179861.3 | NP_850192.1 | 8 - 9 | 928 | 78 | ||
| NM_179862.2 | NP_850193.1 | 7 - 8 | 1057 | 78 | ||
| NM_179863.3 | NP_850194.1 | 8 - 9 | 1024 | 78 | ||
| NM_179864.3 | NP_850195.2 | 8 - 9 | 873 | 78 | ||
| GenBank mRNA | AF367306.1 | - | 8 - 9 | 970 | 78 | |
| BT002219.1 | - | 6 - 7 | 703 | 78 | ||
More Information
Set Membership: |
|

LUH
Genomic Map