Assay Details
| Gene Symbol: AT2G32710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128830.4 | NP_565750.1 | 2 - 3 | 936 | 70 | |
| NM_179865.2 | NP_850196.1 | 2 - 3 | 927 | 70 | ||
| GenBank mRNA | AF123315.1 | - | 2 - 3 | 802 | 70 | |
| AJ301555.1 | - | 2 - 3 | 793 | 70 | ||
| BT033128.1 | - | 2 - 3 | 802 | 70 | ||
More Information
Set Membership: |
|

AT2G32710
Genomic Map