Assay Details
| Gene Symbol: AT2G32730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128832.5 | NP_180832.1 | 5 - 6 | 833 | 66 | |
| GenBank mRNA | AY099861.1 | - | 5 - 6 | 732 | 66 | |
More Information
Set Membership: |
|

AT2G32730
Genomic Map