Assay Details
| Gene Symbol: BGAL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202731.1 | NP_001189660.1 | 5 - 6 | 592 | 82 | |
| NM_128841.5 | NP_565755.1 | 5 - 6 | 662 | 82 | ||
| GenBank mRNA | AJ270305.1 | - | 5 - 6 | 566 | 82 | |
More Information
Set Membership: |
|

BGAL9
Genomic Map