Assay Details
| Gene Symbol: BGAL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202731.1 | NP_001189660.1 | 7 - 8 | 829 | 67 | |
| NM_128841.5 | NP_565755.1 | 7 - 8 | 899 | 67 | ||
| GenBank mRNA | AJ270305.1 | - | 7 - 8 | 803 | 67 | |
More Information
Set Membership: |
|

BGAL9
Genomic Map