Assay Details
| Gene Symbol: BGLU33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128846.4 | NP_180845.2 | 12 - 13 | 1596 | 109 | |
| NM_201858.1 | NP_973587.1 | 11 - 12 | 1599 | 109 | ||
More Information
Set Membership: |
|

BGLU33
Genomic Map