Assay Details
| Gene Symbol: BGLU33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128846.4 | NP_180845.2 | 3 - 4 | 510 | 74 | |
| NM_201858.1 | NP_973587.1 | 3 - 4 | 510 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BGLU33| Gene Symbol: BGLU33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128846.4 | NP_180845.2 | 3 - 4 | 510 | 74 | |
| NM_201858.1 | NP_973587.1 | 3 - 4 | 510 | 74 | ||
Set Membership: |
|