Assay Details
| Gene Symbol: BGLU33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128846.4 | NP_180845.2 | 5 - 6 | 672 | 135 | |
| NM_201858.1 | NP_973587.1 | 5 - 6 | 672 | 135 | ||
More Information
Set Membership: |
|

BGLU33
Genomic Map