Assay Details
| Gene Symbol: ATZW10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128850.3 | NP_565757.2 | 2 - 3 | 838 | 94 | |
| GenBank mRNA | AY069883.1 | - | 2 - 3 | 772 | 94 | |
| U80984.1 | - | 2 - 3 | 760 | 94 | ||
More Information
Set Membership: |
|

ATZW10
Genomic Map