Assay Details
| Gene Symbol: ATZW10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128850.3 | NP_565757.2 | 8 - 9 | 1710 | 114 | |
| GenBank mRNA | AY069883.1 | - | 8 - 9 | 1644 | 114 | |
| U80984.1 | - | 8 - 9 | 1632 | 114 | ||
More Information
Set Membership: |
|

ATZW10
Genomic Map