Assay Details
| Gene Symbol: ATZW10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128850.3 | NP_565757.2 | 9 - 10 | 1835 | 140 | |
| GenBank mRNA | AY069883.1 | - | 9 - 10 | 1769 | 140 | |
| U80984.1 | - | 9 - 10 | 1757 | 140 | ||
More Information
Set Membership: |
|

ATZW10
Genomic Map