Assay Details
| Gene Symbol: COP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336418.1 | NP_001325309.1 | 9 - 10 | 1573 | 71 | |
| NM_128855.4 | NP_180854.1 | 9 - 10 | 1576 | 71 | ||
| GenBank mRNA | BT025337.1 | - | 9 - 10 | 1457 | 71 | |
More Information
Set Membership: |
|

COP1
Genomic Map