Assay Details
| Gene Symbol: PFA-DSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128856.5 | NP_180855.2 | 1 - 2 | 593 | 84 | |
| GenBank mRNA | AK227524.1 | - | 1 - 2 | 495 | 84 | |
| BT006379.1 | - | 1 - 2 | 286 | 84 | ||
More Information
Set Membership: |
|

PFA-DSP2
Genomic Map