Assay Details
| Gene Symbol: PFA-DSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128856.5 | NP_180855.2 | 4 - 5 | 845 | 88 | |
| GenBank mRNA | AK227524.1 | - | 4 - 5 | 747 | 88 | |
| BT006379.1 | - | 4 - 5 | 538 | 88 | ||
More Information
Set Membership: |
|

PFA-DSP2
Genomic Map