Assay Details
| Gene Symbol: ATP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128864.4 | NP_180863.1 | 6 - 7 | 796 | 71 | |
| GenBank mRNA | AY039513.1 | - | 6 - 7 | 744 | 71 | |
| AY062627.1 | - | 6 - 7 | 757 | 71 | ||
| AY086685.1 | - | 6 - 7 | 791 | 71 | ||
| AY102152.1 | - | 6 - 7 | 658 | 71 | ||
| D88374.1 | - | 6 - 7 | 732 | 71 | ||
More Information
Set Membership: |
|

ATP3
Genomic Map