Assay Details
| Gene Symbol: ATP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128864.4 | NP_180863.1 | 7 - 8 | 914 | 61 | |
| GenBank mRNA | AY039513.1 | - | 7 - 8 | 862 | 61 | |
| AY062627.1 | - | 7 - 8 | 875 | 61 | ||
| AY086685.1 | - | 7 - 8 | 909 | 61 | ||
| AY102152.1 | - | 7 - 8 | 776 | 61 | ||
| D88374.1 | - | 7 - 8 | 850 | 61 | ||
More Information
Set Membership: |
|

ATP3
Genomic Map