Assay Details
| Gene Symbol: CSLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336422.1 | NP_001324935.1 | 3 - 4 | 1091 | 75 | |
| NM_128870.4 | NP_180869.1 | 2 - 3 | 1095 | 75 | ||
| GenBank mRNA | KJ138876.1 | - | 2 - 3 | 982 | 75 | |
More Information
Set Membership: |
|

CSLD1
Genomic Map