Assay Details
| Gene Symbol: SUVH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336444.1 | NP_001318341.1 | 2 - 3 | 2180 | 98 | |
| NM_128889.3 | NP_180887.1 | 1 - 2 | 2118 | 98 | ||
| GenBank mRNA | AF344445.1 | - | 1 - 2 | 2118 | 98 | |
More Information
Set Membership: |
|

SUVH2
Genomic Map