Assay Details
| Gene Symbol: CPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336459.1 | NP_001325202.1 | 9 - 10 | 4136 | 99 | |
| NM_128914.3 | NP_180912.2 | 10 - 11 | 4011 | 99 | ||
More Information
Set Membership: |
|

CPL3
Genomic Map