Assay Details
| Gene Symbol: BUBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202739.2 | NP_001189668.1 | 3 - 4 | 545 | 129 | |
| NM_128916.3 | NP_565767.1 | 3 - 4 | 545 | 129 | ||
More Information
Set Membership: |
|

BUBR1
Genomic Map