Assay Details
| Gene Symbol: SWI3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128921.6 | NP_180919.2 | 1 - 2 | 213 | 88 | |
| GenBank mRNA | BT004085.1 | - | 1 - 2 | 195 | 88 | |
| BT005094.1 | - | 1 - 2 | 162 | 88 | ||
More Information
Set Membership: |
|

SWI3B
Genomic Map