Assay Details
| Gene Symbol: HB-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336476.1 | NP_001324695.1 | 1 - 2 | 303 | 87 | |
| NM_128948.4 | NP_180944.2 | 1 - 2 | 321 | 87 | ||
| GenBank mRNA | AY251401.1 | - | 1 - 2 | 269 | 87 | |
More Information
Set Membership: |
|

HB-3
Genomic Map