Assay Details
| Gene Symbol: AT1G51350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104013.3 | NP_175546.1 | 2 - 3 | 1295 | 78 | |
| GenBank mRNA | AY093140.1 | - | 2 - 3 | 1184 | 78 | |
| BT008416.1 | - | 2 - 3 | 1088 | 78 | ||
More Information
Set Membership: |
|

AT1G51350
Genomic Map