Assay Details
| Gene Symbol: TRO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104023.3 | NP_175556.1 | 1 - 2 | 1334 | 133 | |
| GenBank mRNA | AK229176.1 | - | 1 - 2 | 1291 | 133 | |
| BT023430.1 | - | 1 - 2 | 1273 | 133 | ||
More Information
Set Membership: |
|

TRO
Genomic Map