Assay Details
| Gene Symbol: SYP81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333499.1 | NP_001323219.1 | 5 - 6 | 1150 | 67 | |
| NM_104052.3 | NP_564597.1 | 6 - 7 | 1003 | 67 | ||
| GenBank mRNA | BT002917.1 | - | 6 - 7 | 972 | 67 | |
| BT004352.1 | - | 6 - 7 | 783 | 67 | ||
More Information
Set Membership: |
|

SYP81
Genomic Map