Assay Details
| Gene Symbol: MBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333522.1 | NP_001321720.1 | 2 - 3 | 348 | 69 | |
| NM_104084.4 | NP_175615.1 | 2 - 3 | 380 | 69 | ||
| NM_179463.3 | NP_849794.1 | 1 - 2 | 862 | 69 | ||
| GenBank mRNA | AY039537.1 | - | 2 - 3 | 311 | 69 | |
| BT002280.1 | - | 1 - 2 | 193 | 69 | ||
More Information
Set Membership: |
|

MBP2
Genomic Map