Assay Details
| Gene Symbol: AR791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333524.1 | NP_001323019.1 | 4 - 5 | 1092 | 124 | |
| NM_104089.4 | NP_564600.1 | 4 - 5 | 1394 | 124 | ||
| GenBank mRNA | AF367324.1 | - | 4 - 5 | 1139 | 124 | |
| AY133607.1 | - | 3 - 4 | 958 | 124 | ||
More Information
Set Membership: |
|

AR791
Genomic Map