Assay Details
| Gene Symbol: SK21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125404.1 | NP_001118876.1 | 3 - 4 | 532 | 124 | |
| NM_116006.4 | NP_567113.1 | 3 - 4 | 542 | 124 | ||
| GenBank mRNA | AY084971.1 | - | 3 - 4 | 518 | 124 | |
More Information
Set Membership: |
|

SK21
Genomic Map