Assay Details
| Gene Symbol: SK21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125404.1 | NP_001118876.1 | 8 - 9 | 1177 | 153 | |
| NM_116006.4 | NP_567113.1 | 8 - 9 | 1187 | 153 | ||
| GenBank mRNA | AY084971.1 | - | 8 - 9 | 1163 | 153 | |
More Information
Set Membership: |
|

SK21
Genomic Map