Assay Details
| Gene Symbol: SYP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203224.2 | NP_001190153.1 | 2 - 3 | 385 | 87 | |
| NM_001340100.1 | NP_001326526.1 | 2 - 3 | 372 | 87 | ||
| NM_001340101.1 | NP_001326527.1 | 2 - 3 | 372 | 87 | ||
| NM_001340102.1 | NP_001326528.1 | 1 - 2 | 436 | 87 | ||
| NM_001340103.1 | NP_001326529.1 | 2 - 3 | 354 | 87 | ||
| NM_116010.1 | NP_567114.1 | 2 - 3 | 344 | 87 | ||
| GenBank mRNA | AF355759.1 | - | 2 - 3 | 344 | 87 | |
More Information
Set Membership: |
|

SYP73
Genomic Map