Assay Details
| Gene Symbol: SYP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203224.2 | NP_001190153.1 | 3 - 4 | 546 | 86 | |
| NM_001340100.1 | NP_001326526.1 | 3 - 4 | 533 | 86 | ||
| NM_001340101.1 | NP_001326527.1 | 3 - 4 | 533 | 86 | ||
| NM_001340102.1 | NP_001326528.1 | 2 - 3 | 597 | 86 | ||
| NM_001340103.1 | NP_001326529.1 | 3 - 4 | 515 | 86 | ||
| NM_116010.1 | NP_567114.1 | 3 - 4 | 505 | 86 | ||
| GenBank mRNA | AF355759.1 | - | 3 - 4 | 505 | 86 | |
More Information
Set Membership: |
|

SYP73
Genomic Map