Assay Details
| Gene Symbol: SYP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203224.2 | NP_001190153.1 | 5 - 6 | 693 | 75 | |
| NM_001340100.1 | NP_001326526.1 | 5 - 6 | 680 | 75 | ||
| NM_001340101.1 | NP_001326527.1 | 5 - 6 | 680 | 75 | ||
| NM_001340102.1 | NP_001326528.1 | 4 - 5 | 744 | 75 | ||
| NM_001340103.1 | NP_001326529.1 | 5 - 6 | 662 | 75 | ||
| NM_116010.1 | NP_567114.1 | 5 - 6 | 649 | 75 | ||
| GenBank mRNA | AF355759.1 | - | 5 - 6 | 649 | 75 | |
More Information
Set Membership: |
|

SYP73
Genomic Map