Assay Details
| Gene Symbol: SYP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203224.2 | NP_001190153.1 | 8 - 9 | 876 | 68 | |
| NM_001340102.1 | NP_001326528.1 | 7 - 8 | 927 | 68 | ||
| NM_116010.1 | NP_567114.1 | 8 - 9 | 832 | 68 | ||
| GenBank mRNA | AF355759.1 | - | 8 - 9 | 832 | 68 | |
More Information
Set Membership: |
|

SYP73
Genomic Map