Assay Details
| Gene Symbol: DL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340126.1 | NP_001325984.1 | 4 - 5 | 539 | 71 | |
| NM_001340127.1 | NP_001325985.1 | 4 - 5 | 539 | 71 | ||
| NM_116041.3 | NP_191735.2 | 4 - 5 | 539 | 71 | ||
| GenBank mRNA | AY189279.1 | - | 4 - 5 | 463 | 71 | |
More Information
Set Membership: |
|

DL1B
Genomic Map