Assay Details
| Gene Symbol: AT3G62530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116118.4 | NP_567129.1 | 3 - 4 | 632 | 77 | |
| GenBank mRNA | AF370303.1 | - | 3 - 4 | 617 | 77 | |
| AY063090.1 | - | 3 - 4 | 535 | 77 | ||
More Information
Set Membership: |
|

AT3G62530
Genomic Map