Assay Details
| Gene Symbol: DRB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161214.2 | NP_001154686.1 | 3 - 4 | 675 | 80 | |
| NM_116145.4 | NP_191839.2 | 3 - 4 | 682 | 80 | ||
| NM_202751.1 | NP_974480.1 | 3 - 4 | 677 | 80 | ||
| GenBank mRNA | AB455097.1 | - | 2 - 3 | 605 | 80 | |
| AY150509.1 | - | 2 - 3 | 605 | 80 | ||
More Information
Set Membership: |
|

DRB4
Genomic Map