Assay Details
| Gene Symbol: TIR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116163.4 | NP_567135.1 | 1 - 2 | 913 | 67 | |
| GenBank mRNA | AF005048.1 | - | 1 - 2 | 607 | 67 | |
| BT001946.1 | - | 1 - 2 | 464 | 67 | ||
More Information
Set Membership: |
|

TIR1
Genomic Map