Assay Details
| Gene Symbol: TIR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116163.4 | NP_567135.1 | 2 - 3 | 1410 | 70 | |
| GenBank mRNA | AF005048.1 | - | 2 - 3 | 1104 | 70 | |
| BT001946.1 | - | 2 - 3 | 961 | 70 | ||
More Information
Set Membership: |
|

TIR1
Genomic Map