Assay Details
| Gene Symbol: AT3G63070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116172.3 | NP_191866.3 | 3 - 4 | 2545 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G63070| Gene Symbol: AT3G63070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116172.3 | NP_191866.3 | 3 - 4 | 2545 | 67 | |
Set Membership: |
|