Assay Details
| Gene Symbol: MIRO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116180.4 | NP_567139.1 | 4 - 5 | 1059 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIRO2| Gene Symbol: MIRO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116180.4 | NP_567139.1 | 4 - 5 | 1059 | 77 | |
Set Membership: |
|