Assay Details
| Gene Symbol: BIL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116196.3 | NP_191890.1 | 2 - 3 | 510 | 75 | |
| GenBank mRNA | BT020236.1 | - | 2 - 3 | 444 | 75 | |
| BT021132.1 | - | 2 - 3 | 354 | 75 | ||
More Information
Set Membership: |
|

BIL4
Genomic Map