Assay Details
| Gene Symbol: EMB3126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116214.3 | NP_191908.1 | 5 - 6 | 841 | 76 | |
| GenBank mRNA | AY053410.1 | - | 5 - 6 | 829 | 76 | |
| AY059929.1 | - | 5 - 6 | 828 | 76 | ||
| AY133519.1 | - | 5 - 6 | 775 | 76 | ||
| BT001191.1 | - | 5 - 6 | 775 | 76 | ||
More Information
Set Membership: |
|

EMB3126
Genomic Map