Assay Details
| Gene Symbol: MEE50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116241.4 | NP_567156.1 | 1 - 2 | 635 | 73 | |
| GenBank mRNA | AK118272.1 | - | 1 - 2 | 612 | 73 | |
| BT005533.1 | - | 1 - 2 | 583 | 73 | ||
More Information
Set Membership: |
|

MEE50
Genomic Map