Assay Details
| Gene Symbol: GPAT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116264.6 | NP_191950.2 | 2 - 3 | 760 | 77 | |
| GenBank mRNA | BT015813.1 | - | 2 - 3 | 636 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPAT8| Gene Symbol: GPAT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116264.6 | NP_191950.2 | 2 - 3 | 760 | 77 | |
| GenBank mRNA | BT015813.1 | - | 2 - 3 | 636 | 77 | |
Set Membership: |
|