Assay Details
| Gene Symbol: GPAT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116264.6 | NP_191950.2 | 3 - 4 | 1236 | 62 | |
| GenBank mRNA | BT015813.1 | - | 3 - 4 | 1112 | 62 | |
More Information
Set Membership: |
|

GPAT8
Genomic Map